siRNA Details
Gene Symbol: CFAP206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031743.2 | NP_001026913.1 | 9 | 1123 | ||
GenBank mRNA | AK093830.1 | - | 9 | 1082 | ||
AK098840.1 | - | 3 | 350 | |||
BC035083.2 | - | 9 | 1092 | 5260875 |
