siRNA Details
Gene Symbol: CFAP206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031743.2 | NP_001026913.1 | 5 | 436 | ||
GenBank mRNA | AK093830.1 | - | 5 | 395 | ||
BC035083.2 | - | 5 | 405 | 5260875 | ||
DB021694.1 | - | 5 | 432 |
