siRNA Details
Gene Symbol: ZNF782 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001662.1 | NP_001001662.1 | 6 | 972 | ||
XM_005251742.4 | XP_005251799.1 | 5 | 991 | |||
XM_005251744.3 | XP_005251801.1 | 4 | 932 | |||
XM_011518315.2 | XP_011516617.1 | 6 | 2183 | |||
XM_011518318.2 | XP_011516620.1 | 6 | 1170 | |||
XM_011518320.2 | XP_011516622.1 | 5 | 807 | |||
XM_017014343.1 | XP_016869832.1 | 5 | 2359 | |||
XM_017014344.1 | XP_016869833.1 | 4 | 704 | |||
GenBank mRNA | AK131468.1 | - | 6 | 972 | ||
AK302460.1 | - | 3 | 656 | |||
BC137073.1 | - | 6 | 969 | 9021070 |
