siRNA Details
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020945.1 | NP_065996.1 | 10, 11 | 1752 | ||
XM_005270004.3 | XP_005270061.1 | 10, 11 | 1867 | |||
XM_011539986.2 | XP_011538288.2 | 10, 11 | 1870 | |||
XM_011539987.2 | XP_011538289.2 | 10, 11 | 1873 | |||
XM_011539988.2 | XP_011538290.1 | 11, 12 | 1951 | |||
XM_011539990.2 | XP_011538292.1 | 10, 11 | 1898 | |||
XM_011539991.2 | XP_011538293.1 | 10, 11 | 1902 | |||
XM_011539992.2 | XP_011538294.1 | 10, 11 | 1902 | |||
XM_017016463.1 | XP_016871952.1 | 11, 12 | 1794 | |||
XM_017016465.1 | XP_016871954.1 | 10, 11 | 1879 | |||
GenBank mRNA | AK127650.1 | - | 10, 11 | 1761 | ||
BC034937.1 | - | 10, 11 | 1818 |
