siRNA Details
Gene Symbol: DEUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181645.3 | NP_857596.2 | 4, 5 | 386 | ||
XM_005273802.1 | XP_005273859.1 | 4, 5 | 433 | |||
XM_011542629.2 | XP_011540931.1 | 4, 5 | 416 | |||
XM_011542630.1 | XP_011540932.1 | 4, 5 | 433 | |||
XM_011542631.2 | XP_011540933.1 | 4, 5 | 1493 | |||
XM_011542632.2 | XP_011540934.1 | 4, 5 | 416 | |||
XM_011542633.2 | XP_011540935.1 | 4, 5 | 416 | |||
XM_011542635.2 | XP_011540937.1 | 4, 5 | 416 | |||
XM_011542637.2 | XP_011540939.1 | 4, 5 | 416 | |||
XM_017017245.1 | XP_016872734.1 | 4, 5 | 416 | |||
GenBank mRNA | AK058122.1 | - | 2, 3 | 120 | ||
AK127973.1 | - | 4, 5 | 433 | |||
BC017929.1 | - | 4, 5 | 365 |
