siRNA Details
Gene Symbol: TMTC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181783.3 | NP_861448.2 | 7 | 1221 | ||
XM_011537980.2 | XP_011536282.1 | 6 | 996 | |||
GenBank mRNA | AJ697717.1 | - | 7 | 1091 | ||
AK074973.1 | - | 6 | 848 | |||
AK290436.1 | - | 7 | 1221 | |||
AK302755.1 | - | 7 | 1221 | |||
AK310375.1 | - | 7 | 1221 | |||
AK310393.1 | - | 7 | 1221 | |||
BC117175.1 | - | 6 | 1042 | 40125726 | ||
BC117177.1 | - | 6 | 1043 | 40125728 | ||
BX538169.1 | - | 7 | 1194 | |||
CR749244.1 | - | 7 | 1173 | |||
CR749309.1 | - | 7 | 1207 | |||
CR933632.1 | - | 7 | 1172 |
