siRNA Details
Gene Symbol: ZNF169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301275.1 | NP_001288204.1 | 5 | 1011 | ||
NM_003448.2 | NP_003439.2 | 5 | 647 | |||
NM_194320.3 | NP_919301.2 | 5 | 644 | |||
XM_017014362.1 | XP_016869851.1 | 5 | 776 | |||
XM_017014363.1 | XP_016869852.1 | 6 | 670 | |||
XM_017014364.1 | XP_016869853.1 | 6 | 667 | |||
XM_017014365.1 | XP_016869854.1 | 5 | 7000 | |||
XM_017014366.1 | XP_016869855.1 | 5 | 6938 | |||
GenBank mRNA | AK093956.1 | - | 5 | 949 | ||
AK122832.1 | - | 1 | 1267 | |||
AK289742.1 | - | 5 | 588 | |||
BC047702.2 | - | 4 | 568 | 5585128 |
