siRNA Details
Gene Symbol: ZNF169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301275.1 | NP_001288204.1 | 5 | 1140 | ||
NM_003448.2 | NP_003439.2 | 5 | 776 | |||
NM_194320.3 | NP_919301.2 | 5 | 773 | |||
XM_017014362.1 | XP_016869851.1 | 5 | 905 | |||
XM_017014363.1 | XP_016869852.1 | 6 | 799 | |||
XM_017014364.1 | XP_016869853.1 | 6 | 796 | |||
XM_017014365.1 | XP_016869854.1 | 5 | 7129 | |||
XM_017014366.1 | XP_016869855.1 | 5 | 7067 | |||
GenBank mRNA | AK093956.1 | - | 5 | 1078 | ||
AK122832.1 | - | 1 | 1396 | |||
AK289742.1 | - | 5 | 717 | |||
BC047702.2 | - | 4 | 697 | 5585128 |
