siRNA Details
Gene Symbol: PSORS1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014068.2 | NP_054787.2 | 4, 5 | 323 | ||
GenBank mRNA | AB031479.1 | - | 4, 5 | 308 | ||
AF484418.1 | - | 4, 5 | 324 | |||
DB454051.1 | - | 2, 3 | 110 |
