siRNA Details
Gene Symbol: ADAMTSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040272.5 | NP_001035362.3 | 22 | 4163 | ||
XM_011518063.2 | XP_011516365.1 | 24 | 4301 | |||
XM_011518064.2 | XP_011516366.1 | 23 | 4416 | |||
XM_011518067.1 | XP_011516369.1 | 14 | 3031 | |||
XM_011518068.2 | XP_011516370.1 | 12 | 3634 | |||
XM_011518070.2 | XP_011516372.1 | 12 | 2799 | |||
XM_017015310.1 | XP_016870799.1 | 23 | 10938 | |||
XM_017015311.1 | XP_016870800.1 | 23 | 4250 | |||
XM_017015312.1 | XP_016870801.1 | 22 | 4366 | |||
XM_017015313.1 | XP_016870802.1 | 23 | 4064 | |||
XM_017015314.1 | XP_016870803.1 | 24 | 4301 | |||
GenBank mRNA | AK092602.1 | - | 1 | 222 | ||
BC100788.2 | - | 1 | 212 | |||
BC100789.2 | - | 1 | 214 | |||
BC100790.2 | - | 1 | 211 | |||
BX640951.1 | - | 3 | 296 |
