siRNA Details
Gene Symbol: ZNF804B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181646.3 | NP_857597.1 | 4 | 1167 | ||
GenBank mRNA | AK056672.1 | - | 4 | 1096 | ||
AY295769.1 | - | 4 | 1096 | |||
BC140826.1 | - | 4 | 1167 | 9021694 | ||
BC140829.1 | - | 4 | 1167 | 9021697 |
