siRNA Details
Gene Symbol: C6orf136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109938.2 | NP_001103408.1 | 5 | 1012 | ||
NM_001161376.1 | NP_001154848.1 | 5 | 1555 | |||
NM_145029.3 | NP_659466.2 | 4 | 610 | |||
XM_011514384.1 | XP_011512686.1 | 6 | 1104 | |||
GenBank mRNA | AK127564.1 | - | 5 | 972 | ||
BC016167.1 | - | 5 | 1163 | 3509511 | ||
BC064397.1 | - | 5 | 1173 | |||
BM695485.1 | - | 4 | 485 | |||
BQ022831.1 | - | 2 | 432 |
