siRNA Details
Gene Symbol: LACE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323005.1 | NP_001309934.1 | 3 | 606 | ||
NM_145315.4 | NP_660358.2 | 3 | 606 | |||
XM_005266885.3 | XP_005266942.1 | 4 | 1964 | |||
XM_011535657.2 | XP_011533959.1 | 3 | 489 | |||
XM_011535659.2 | XP_011533961.1 | 3 | 489 | |||
XM_011535660.2 | XP_011533962.1 | 3 | 489 | |||
XM_011535661.2 | XP_011533963.1 | 3 | 489 | |||
XM_017010647.1 | XP_016866136.1 | 4 | 456 | |||
XM_017010648.1 | XP_016866137.1 | 2 | 58 | |||
GenBank mRNA | AF520418.1 | - | 3 | 586 | ||
BC018445.1 | - | 3 | 586 | 3871205 |
