siRNA Details
Gene Symbol: RNF214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077239.1 | NP_001070707.1 | 14 | 2010 | ||
NM_001278249.1 | NP_001265178.1 | 14 | 1545 | |||
NM_207343.3 | NP_997226.2 | 14 | 2072 | |||
GenBank mRNA | AK298923.1 | - | 14 | 2004 | ||
AK300160.1 | - | 14 | 1545 | |||
AL834448.1 | - | 4 | 552 | |||
BC031347.1 | - | 9 | 1107 | |||
BC146891.1 | - | 14 | 2010 | 9056819 | ||
BC146901.1 | - | 14 | 2010 | 9056829 |
