siRNA Details
Gene Symbol: MRGPRG-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027138.1 | 2 | 678 | |||
GenBank mRNA | AK093421.1 | - | 2 | 678 | ||
AK097749.1 | - | 3 | 711 | |||
AY422795.1 | - | 1 | 345 | |||
BC104971.1 | - | 1 | 383 | |||
BC104973.1 | - | 1 | 383 |
