siRNA Details
Gene Symbol: MYO1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101421.3 | NP_001094891.3 | 27 | 2701 | ||
XM_011538223.2 | XP_011536525.1 | 28 | 3209 | |||
XM_017019207.1 | XP_016874696.1 | 27 | 3143 | |||
GenBank mRNA | AK094906.1 | - | 4 | 472 | ||
AK298093.1 | - | 5 | 549 | |||
DC359165.1 | - | 5 | 550 | |||
DC401393.1 | - | 1 | 13 |
