siRNA Details
Gene Symbol: GOLGA6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304388.1 | NP_001291317.1 | 8 | 2779 | ||
GenBank mRNA | AK093463.1 | - | 9 | 1800 | ||
AK097414.1 | - | 1506 | ||||
AK128822.1 | - | 1468 | ||||
BC129820.1 | - | 9 | 1119 | 40113145 | ||
BC129821.1 | - | 8 | 1715 |
