siRNA Details
Gene Symbol: PRR19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199285.2 | NP_954979.2 | 2 | 636 | ||
XM_005258779.4 | XP_005258836.2 | 2 | 574 | |||
XM_005258780.3 | XP_005258837.1 | 2 | 371 | |||
XM_006723154.3 | XP_006723217.1 | 2 | 519 | |||
XM_011526787.1 | XP_011525089.1 | 2 | 337 | |||
XM_011526788.1 | XP_011525090.1 | 2 | 348 | |||
XM_011526789.2 | XP_011525091.1 | 3 | 340 | |||
XM_011526790.2 | XP_011525092.1 | 2 | 359 | |||
GenBank mRNA | AK124116.1 | - | 1 | 1069 | ||
AK291528.1 | - | 2 | 637 | |||
BC063021.1 | - | 2 | 670 | 5245434 |
