siRNA Details
Gene Symbol: FLJ33534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040080.1 | 3 | 412 | |||
GenBank mRNA | AK090853.1 | - | 2 | 359 | ||
AK311163.1 | - | 4 | 874 | |||
BC111723.1 | - | 3 | 412 | 40024134 |
