siRNA Details
Gene Symbol: ZNF619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145082.2 | NP_001138554.1 | 3 | 468 | ||
NM_001145093.2 | NP_001138565.1 | 3 | 468 | |||
NR_026746.2 | 3 | 468 | ||||
XM_006713112.2 | XP_006713175.1 | 3 | 439 | |||
XM_011533607.1 | XP_011531909.1 | 3 | 439 | |||
XM_011533608.1 | XP_011531910.1 | 3 | 439 | |||
XM_017006225.1 | XP_016861714.1 | 2 | 363 | |||
GenBank mRNA | AK075245.1 | - | 3 | 435 | ||
AK304741.1 | - | 3 | 397 | |||
BC117169.1 | - | 1 | 149 | 40125720 | ||
BC143304.1 | - | 1 | 149 | |||
BC143306.1 | - | 1 | 149 | 9051811 | ||
BC143331.1 | - | 1 | 149 | |||
DC308744.1 | - | 3 | 468 |
