siRNA Details
Gene Symbol: LSMEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134468.1 | NP_001127940.1 | 2 | 199 | ||
NM_182597.2 | NP_872403.1 | 2 | 405 | |||
XM_011516074.1 | XP_011514376.1 | 2 | 908 | |||
XM_011516075.2 | XP_011514377.1 | 2 | 2595 | |||
XM_011516076.2 | XP_011514378.1 | 2 | 2231 | |||
XM_017012028.1 | XP_016867517.1 | 1 | 2113 | |||
GenBank mRNA | AK096894.1 | - | 2 | 421 | ||
BC031976.1 | - | 3 | 383 | |||
BC126149.1 | - | 2 | 243 | 8991865 | ||
BC130568.1 | - | 2 | 243 | 40146599 | ||
DA894907.1 | - | 2 | 390 | |||
DA898266.1 | - | 2 | 401 | |||
DB456503.2 | - | 2 | 199 |
