siRNA Details
Gene Symbol: FAM151A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176782.2 | NP_788954.2 | 5 | 798 | ||
XM_006710599.3 | XP_006710662.1 | 6 | 874 | |||
GenBank mRNA | AK091901.1 | - | 5 | 827 | ||
AY358629.1 | - | 5 | 784 | |||
BC015993.1 | - | 5 | 800 | 4614435 | ||
BC020874.1 | - | 5 | 796 | 4608844 |
