siRNA Details
Gene Symbol: KY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178554.4 | NP_848649.3 | 11 | 1253 | ||
XM_005247418.3 | XP_005247475.1 | 10 | 6221 | |||
XM_006713612.3 | XP_006713675.1 | 10 | 6167 | |||
XM_017006287.1 | XP_016861776.1 | 11 | 6299 | |||
XM_017006288.1 | XP_016861777.1 | 11 | 6245 | |||
XM_017006289.1 | XP_016861778.1 | 10 | 6236 | |||
XM_017006290.1 | XP_016861779.1 | 10 | 6182 | |||
GenBank mRNA | AK090526.1 | - | 11 | 1253 | ||
AK293840.1 | - | 10 | 1186 | |||
BC136349.1 | - | 10 | 1023 | |||
DA212239.1 | - | 3 | 463 |
