siRNA Details
Gene Symbol: MS4A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206893.3 | NP_996776.2 | 3 | 364 | ||
XM_011544989.1 | XP_011543291.1 | 3 | 364 | |||
XM_011544991.2 | XP_011543293.1 | 2 | 175 | |||
GenBank mRNA | AK122633.1 | - | 3 | 365 | ||
BC137259.1 | - | 3 | 314 | 9021256 | ||
BC137263.1 | - | 3 | 314 | 9021260 |
