siRNA Details
Gene Symbol: OR6C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005494.1 | NP_001005494.1 | 1 | 848 | ||
GenBank mRNA | BC137000.1 | - | 1 | 876 | 9020992 | |
BC137001.1 | - | 1 | 876 | 9020993 |
