siRNA Details
Gene Symbol: ZNF677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317998.1 | NP_001304927.1 | 4, 5 | 544 | ||
NM_182609.3 | NP_872415.1 | 4, 5 | 362 | |||
XM_011526912.2 | XP_011525214.1 | 4, 5 | 490 | |||
GenBank mRNA | AK308120.1 | - | 4, 5 | 324 | ||
BC029855.1 | - | 4, 5 | 351 | |||
BC050038.1 | - | 4, 5 | 317 | 5272569 | ||
BX648243.1 | - | 4, 5 | 333 | |||
CK125988.1 | - | 4, 5 | 548 |
