siRNA Details
Gene Symbol: EYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142800.1 | NP_001136272.1 | 10 | 2083 | ||
NM_001142801.1 | NP_001136273.1 | 10 | 2083 | |||
NM_001292009.1 | NP_001278938.1 | 10 | 2083 | |||
NM_198283.1 | NP_938024.1 | 9 | 1942 | |||
GenBank mRNA | AB593114.1 | - | 10 | 2048 | ||
AY358133.1 | - | 9 | 1942 | |||
BC133011.1 | - | 8 | 1807 | |||
BC133013.1 | - | 8 | 1804 | |||
BX537914.1 | - | 10 | 2083 | |||
FJ416331.1 | - | 10 | 2091 | |||
FM209056.1 | - | 10 | 2035 |
