siRNA Details
Gene Symbol: EYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142800.1 | NP_001136272.1 | 4 | 1070 | ||
NM_001142801.1 | NP_001136273.1 | 4 | 1070 | |||
NM_001292009.1 | NP_001278938.1 | 4 | 1070 | |||
NM_198283.1 | NP_938024.1 | 3 | 929 | |||
GenBank mRNA | AB593112.1 | - | 4 | 1026 | ||
AB593114.1 | - | 4 | 1035 | |||
AK310478.1 | - | 6 | 1102 | |||
AY358133.1 | - | 3 | 929 | |||
BC133011.1 | - | 2 | 794 | |||
BC133013.1 | - | 2 | 791 | |||
BX537914.1 | - | 4 | 1070 | |||
BX647741.1 | - | 4 | 1060 | |||
FJ416331.1 | - | 4 | 1078 | |||
FM209056.1 | - | 4 | 1022 |
