siRNA Details
Gene Symbol: LANCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170331.1 | NP_001163802.1 | 4 | 1251 | ||
NM_198511.2 | NP_940913.1 | 4 | 1251 | |||
XM_011543904.2 | XP_011542206.1 | 4 | 736 | |||
GenBank mRNA | AK124915.1 | - | 4 | 1251 | ||
BC093667.1 | - | 4 | 1043 | 7939512 | ||
BC093669.1 | - | 4 | 1043 | 7939514 | ||
BC143528.1 | - | 4 | 1043 |
