siRNA Details
Gene Symbol: GALNT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198516.2 | NP_940918.2 | 10 | 2074 | ||
XM_006718224.2 | XP_006718287.1 | 9 | 1919 | |||
XM_011520068.2 | XP_011518370.1 | 10 | 1650 | |||
GenBank mRNA | AB078147.1 | - | 10 | 1653 | ||
AJ626724.1 | - | 10 | 1653 | |||
AK055111.1 | - | 10 | 2056 | |||
BC037341.2 | - | 10 | 2063 | 5265475 | ||
BC060864.1 | - | 10 | 2065 | 30341070 |
