siRNA Details
Gene Symbol: C12orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099336.2 | NP_001092806.1 | 6 | 1292 | ||
NM_001278419.1 | NP_001265348.1 | 7 | 1387 | |||
NM_001278420.1 | NP_001265349.1 | 7 | 1252 | |||
NM_198521.3 | NP_940923.2 | 6 | 1157 | |||
XM_006719377.3 | XP_006719440.1 | 5 | 1095 | |||
XM_011538291.1 | XP_011536593.1 | 6 | 1152 | |||
XM_011538292.2 | XP_011536594.1 | 6 | 1121 | |||
XM_011538293.2 | XP_011536595.1 | 6 | 1128 | |||
XM_011538294.2 | XP_011536596.1 | 6 | 1102 | |||
XM_011538295.1 | XP_011536597.1 | 6 | 1184 | |||
XM_011538297.2 | XP_011536599.1 | 7 | 2168 | |||
XM_011538300.1 | XP_011536602.1 | 7 | 1337 | |||
XM_011538305.1 | XP_011536607.1 | 6 | 1354 | |||
XM_011538306.1 | XP_011536608.1 | 5 | 780 | |||
XM_017019276.1 | XP_016874765.1 | 6 | 1246 | |||
XM_017019277.1 | XP_016874766.1 | 9 | 1621 | |||
XM_017019278.1 | XP_016874767.1 | 8 | 1400 | |||
XM_017019279.1 | XP_016874768.1 | 9 | 1926 | |||
XM_017019280.1 | XP_016874769.1 | 9 | 1626 | |||
GenBank mRNA | AA992269.1 | - | 1 | 214 | ||
AK058052.1 | - | 6 | 1115 | |||
BC044617.1 | - | 7 | 1348 | 5267756 | ||
BC117392.1 | - | 6 | 1115 | 40125943 | ||
BC117394.1 | - | 6 | 1115 | 40125945 | ||
HY193513.1 | - | 1 | 232 |
