siRNA Details
Gene Symbol: ZNF470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001668.3 | NP_001001668.3 | 3 | 698 | ||
XM_017026801.1 | XP_016882290.1 | 3 | 698 | |||
GenBank mRNA | AY484591.1 | - | 3 | 214 | ||
BC136762.1 | - | 3 | 210 | 9020752 | ||
BC142627.1 | - | 3 | 693 | |||
BC150188.1 | - | 3 | 532 |
