siRNA Details
Gene Symbol: CFAP77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207417.2 | NP_997300.1 | 2 | 299 | ||
GenBank mRNA | AK128819.1 | - | 2 | 271 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CFAP77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207417.2 | NP_997300.1 | 2 | 299 | ||
GenBank mRNA | AK128819.1 | - | 2 | 271 |