siRNA Details
Gene Symbol: OR51Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004757.2 | NP_001004757.1 | 1 | 816 | ||
GenBank mRNA | BC136999.1 | - | 1 | 816 | 9020991 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR51Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004757.2 | NP_001004757.1 | 1 | 816 | ||
GenBank mRNA | BC136999.1 | - | 1 | 816 | 9020991 |