siRNA Details
Gene Symbol: OR52D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005163.2 | NP_001005163.1 | 1 | 928 | ||
GenBank mRNA | BC136928.1 | - | 1 | 928 | 9020920 | |
BC136930.1 | - | 1 | 928 | 9020922 |
