siRNA Details
Gene Symbol: OR52N4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005175.3 | NP_001005175.3 | 1 | 156 | ||
XM_017017711.1 | XP_016873200.1 | 5 | 4704 | |||
XM_017017712.1 | XP_016873201.1 | 7 | 4864 | |||
XM_017017713.1 | XP_016873202.1 | 4 | 4673 | |||
GenBank mRNA | BC137035.1 | - | 1 | 156 | 9021031 | |
BC137036.1 | - | 1 | 156 | 9021032 |
