siRNA Details
Gene Symbol: OR5AU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004731.1 | NP_001004731.1 | 1 | 974 | ||
GenBank mRNA | BC137304.1 | - | 1 | 1012 | 9021301 | |
BC137305.1 | - | 1 | 1012 | 9021302 |
