siRNA Details
Gene Symbol: MYO18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078471.3 | NP_510880.2 | 31 | 4908 | ||
NM_203318.1 | NP_976063.1 | 31 | 4908 | |||
GenBank mRNA | AB177858.1 | - | 30 | 4727 | ||
AB177860.1 | - | 31 | 3860 | |||
AK302505.1 | - | 30 | 3634 | |||
AK302725.1 | - | 15 | 1622 | |||
AY703984.1 | - | 31 | 3860 | |||
CR933614.1 | - | 31 | 4883 | |||
D86970.2 | - | 31 | 4931 |
