siRNA Details
Gene Symbol: FAM174B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207446.2 | NP_997329.2 | 2 | 554 | ||
GenBank mRNA | AK075564.1 | - | 2 | 548 | ||
BC045729.1 | - | 2 | 130 | |||
BC060873.1 | - | 2 | 445 | 30348143 | ||
BC110328.1 | - | 3 | 282 |
