siRNA Details
Gene Symbol: LINC00898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033377.1 | 3, 4 | 2473 | |||
GenBank mRNA | AK128136.1 | - | 3, 4 | 2470 | ||
BC132762.1 | - | 3, 4 | 806 | |||
BC132788.1 | - | 3, 4 | 806 | |||
BC144501.1 | - | 2, 3 | 725 | |||
BC144502.1 | - | 2, 3 | 854 |
