siRNA Details
Gene Symbol: AMTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286731.1 | NP_001273660.1 | 8 | 671 | ||
NM_212557.3 | NP_997722.1 | 8 | 674 | |||
GenBank mRNA | AB593161.1 | - | 8 | 673 | ||
AY358528.1 | - | 8 | 665 | |||
BC121812.1 | - | 8 | 642 | 40109020 | ||
BC121817.1 | - | 8 | 645 | 40109016 | ||
BU727294.1 | - | 2 | 346 |
