siRNA Details
Gene Symbol: RAB44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257357.1 | NP_001244286.1 | 4 | 524 | ||
XM_011514600.2 | XP_011512902.1 | 4 | 567 | |||
XM_011514601.2 | XP_011512903.1 | 5 | 614 | |||
XM_011514602.2 | XP_011512904.1 | 4 | 567 | |||
XM_011514603.2 | XP_011512905.1 | 4 | 567 | |||
XM_011514604.2 | XP_011512906.1 | 4 | 567 | |||
XM_011514605.2 | XP_011512907.1 | 4 | 567 | |||
XM_011514606.2 | XP_011512908.1 | 4 | 567 | |||
XM_017010855.1 | XP_016866344.1 | 4 | 2006 | |||
GenBank mRNA | AK125083.1 | - | 4 | 524 |
