siRNA Details
Gene Symbol: LINC00238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024338.2 | 5 | 502 | |||
NR_110307.1 | 4 | 349 | ||||
NR_110308.1 | 5 | 502 | ||||
GenBank mRNA | BC033546.1 | - | 5 | 473 | ||
BC056671.1 | - | 5 | 506 | |||
BC070365.1 | - | 5 | 482 | |||
BI829888.1 | - | 4 | 313 |
