siRNA Details
Gene Symbol: OR2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005226.2 | NP_001005226.1 | 1 | 216 | ||
GenBank mRNA | BC136817.1 | - | 1 | 175 | ||
BC136818.1 | - | 1 | 175 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005226.2 | NP_001005226.1 | 1 | 216 | ||
GenBank mRNA | BC136817.1 | - | 1 | 175 | ||
BC136818.1 | - | 1 | 175 |