siRNA Details
Gene Symbol: OR2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005480.2 | NP_001005480.2 | 1 | 927 | ||
GenBank mRNA | BC136742.1 | - | 1 | 996 | 9020732 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005480.2 | NP_001005480.2 | 1 | 927 | ||
GenBank mRNA | BC136742.1 | - | 1 | 996 | 9020732 |