siRNA Details
Gene Symbol: TM4SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014220.2 | NP_055035.1 | 4, 5 | 829 | ||
GenBank mRNA | AK130073.1 | - | 4, 5 | 741 | ||
AK130271.1 | - | 4, 5 | 823 | |||
BC008442.1 | - | 4, 5 | 696 | 4101110 | ||
BC010166.2 | - | 4, 5 | 645 | 4040623 | ||
M90657.1 | - | 703 |
