siRNA Details
Gene Symbol: SSX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278691.1 | NP_001265620.1 | 9 | 811 | ||
NM_005635.3 | NP_005626.1 | 8 | 725 | |||
GenBank mRNA | BC001003.2 | - | 8 | 670 | 3445470 | |
BC125151.1 | - | 8 | 668 | 40120554 | ||
BC128611.1 | - | 8 | 667 | 6159943 | ||
BC133693.1 | - | 8 | 668 | 40120550 | ||
BC150487.2 | - | 8 | 672 | 40120555 | ||
BQ774004.1 | - | 1 | 603 | |||
X86174.1 | - | 8 | 680 |
