siRNA Details
Gene Symbol: SSX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278691.1 | NP_001265620.1 | 9 | 907 | ||
NM_005635.3 | NP_005626.1 | 8 | 821 | |||
GenBank mRNA | BC001003.2 | - | 8 | 766 | 3445470 | |
BC125151.1 | - | 8 | 764 | 40120554 | ||
BC128611.1 | - | 8 | 763 | 6159943 | ||
BC133693.1 | - | 8 | 764 | 40120550 | ||
BC150487.2 | - | 8 | 768 | 40120555 | ||
BQ774004.1 | - | 1 | 507 |
