siRNA Details
Gene Symbol: ZNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164530.1 | NP_001158002.1 | 4 | 520 | ||
NM_001289401.1 | NP_001276330.1 | 5 | 587 | |||
NM_001289402.1 | NP_001276331.1 | 5 | 950 | |||
NM_003436.3 | NP_003427.3 | 5 | 618 | |||
NM_007134.1 | NP_009065.1 | 4 | 556 | |||
XM_005259211.1 | XP_005259268.1 | 4 | 520 | |||
XM_006723362.3 | XP_006723425.1 | 5 | 626 | |||
XM_006723363.3 | XP_006723426.1 | 5 | 990 | |||
XM_017027240.1 | XP_016882729.1 | 4 | 943 | |||
XM_017027241.1 | XP_016882730.1 | 5 | 945 | |||
XM_017027242.1 | XP_016882731.1 | 4 | 385 | |||
GenBank mRNA | AK094202.1 | - | 5 | 909 | ||
AK098011.1 | - | 4 | 520 | |||
AK295110.1 | - | 5 | 588 | |||
AK297226.1 | - | 4 | 351 | |||
BC046434.1 | - | 5 | 582 | 4824328 | ||
BX647775.1 | - | 4 | 524 | |||
DB117846.1 | - | 4 | 520 |
