siRNA Details
Gene Symbol: SNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003498.5 | NP_003489.1 | 2 | 231 | ||
XM_017023741.1 | XP_016879230.1 | 2 | 259 | |||
XM_017023742.1 | XP_016879231.1 | 2 | 157 | |||
GenBank mRNA | AF030196.1 | - | 2 | 183 | ||
AL161976.1 | - | 2 | 192 | |||
BC036100.1 | - | 2 | 209 | 5277179 | ||
BC036443.1 | - | 2 | 203 | 4823944 |
